Learn More
AdipoGen Pterostilbene
SDP
Supplier:  AdipoGen CDXP0234M100
Encompass_Preferred
Chemical. CAS 537-42-8. Formula C16H16O3. MW 256.3. Synthetic. Cell permeable natural methoxylated analog of resveratrol. Antioxidant, antiproliferative, anti-inflammatory, anti-hyperglycemic and anti-diabetic agent. Induces apoptosis. Induces Autophagy. Pterostilbene is a naturally-occurring dimethyl ether analog of resveratrol that is abundant in blueberries. Like resveratrol, pterostilbene acts as a powerful antioxidant, suppresses the synthesis of prostaglandin E2 from lipopolysaccharide-stimulated human peripheral blood mononuclear cells IC50 = 1.0 µM for pterostilbene, 3.2 µM for resveratrol, and inhibits cell proliferation IC50 = ~60 µM for both compounds. Very potent inhibitor of cytochrome P450 1A1 CYP1A1 with Ki=0.57µM. Pterostilbene blocks the activation of ERK1/2, p38 MAPK, and PI3K/Akt signaling pathways, reducing NF-kappaB and AP-1 transcriptional activation. Moderately inhibits COX-1 and COX-2 IC50=19.8µM and 83.9µM.
Catalog No. 50-256-0199