Learn More
Medchemexpress LLC HY-N2157 5mg Medchemexpress, Pteryxin CAS:13161-75-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21575MG
Encompass_Preferred
Medchemexpress, HY-N2157 5mg Pteryxin CAS:13161-75-6 Pteryxin, a coumarin in Peucedanum japonicum Thunb leaves, exerts antiobesity activity. Pteryxin is a potent butyrylcholinesterase (BChE) inhibitor, with an IC50 of 12.96 μg/ml. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2869