Learn More
Cayman Chemical Pteryxn 10mg
SDP
Catalog No. 502992427
Encompass_Preferred
A flavonoid glycoside with diverse biological activities potentiates TRAIL-induced cytotoxicity and apoptosis in U87MG human glioma cells potentiates TRAIL-induced tumor cell apoptosis and reduction of tumor growth in a U87MG mouse xenograft model reduces pulmonary platelet count edema and macrophage polymorphonuclear leukocyte and lymphocyte infiltration as well as inhibits TXNIP/NLRP3 MAPK and NF-KB signaling in a mouse model of LPS-induced acute lung injury from 12 5-50 mg/kg inhibits AChE and induces dyskinesia recovery in a zebrafish model of Alzheimers disease in tank water at 50 g/ml
Catalog No. 50-299-2427 Supplier Cayman Chemical Supplier No. 3032010 MG
A flavonoid glycoside with diverse biological activities potentiates TRAIL-induced cytotoxicity and apoptosis in U87MG human glioma cells potentiates TRAIL-induced tumor cell apoptosis and reduction of tumor growth in a U87MG mouse xenograft model reduces pulmonary platelet count edema and macrophage polymorphonuclear leukocyte and lymphocyte infiltration as well as inhibits TXNIP/NLRP3 MAPK and NF-KB signaling in a mouse model of LPS-induced acute lung injury from 12 5-50 mg/kg inhibits AChE and induces dyskinesia recovery in a zebrafish model of Alzheimers disease in tank water at 50 g/ml