Learn More
IW Tremont Co Inc LabExact Hydrophobic PTFE membrane, NON-STERILE, 0.45 micron, 47mm diameter, pack of 100
SDP
Catalog No. 502237610
Encompass_Preferred
LabExact Polytetrafluoroethylene (PTFE) Membrane is supported with a polypropylene (PP) scrim for easier handling. This scrim is bonded using a unique technique utilizing no bonding agent for purity, chemical compatibility and lower extractables. Hydrophobic material is effective in separating polar/nonpolar solvents. Aqueous solutions can pass after prewetting with methanol
This item is not returnable. View return policy
Catalog No. 50-223-7610 Supplier IW Tremont Co Inc Supplier No. LEIWTPTFE045047
LabExact Polytetrafluoroethylene (PTFE) Membrane is supported with a polypropylene (PP) scrim for easier handling. This scrim is bonded using a unique technique utilizing no bonding agent for purity, chemical compatibility and lower extractables. Hydrophobic material is effective in separating polar/nonpolar solvents. Aqueous solutions can pass after prewetting with methanol