Learn More
CELLTREAT Scientific Products PTFE Syringe Filter, 0.45m, 13mm, Bulk Packed, Non- Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229779
Encompass_Preferred
CELLTREAT Scientific Prodcuts 0.45¿m PTFE Syringe Filters. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. Convenient bulk packaging. PTFE (Polytetrafluoroethylene). Hydrophobic membrane has great temperature resistance. Degassing/clarifying aqueous samples. Strong acid solvent filtration. Alkali solvent filtration. Membrane diameter 13mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, bulk packed, 100/case.
Catalog No. 50-607-522