Learn More
Braintree Scientific PTFE-015 Heat Shrink? Tubing, 1 ft. lengths .038 .018
SDP
Catalog No. 501955585
Encompass
PTFE Heat Shrink Tubing (USP Class VI Approved) Medical grade Polyetrafluoroethylene tubing has high tensile strength and preserves high fidelity pressure waveforms with very small internal diameter tubing. PTFE Heat Shrink tubing can be shrink-fitted to make secure connections with other devices. 2-1 heat-shrink ratio. 2012 Zeus Industrial Products. All rights reserved. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5585 Supplier Braintree Scientific Supplier No. PTFE018
PTFE Heat Shrink Tubing (USP Class VI Approved) Medical grade Polyetrafluoroethylene tubing has high tensile strength and preserves high fidelity pressure waveforms with very small internal diameter tubing. PTFE Heat Shrink tubing can be shrink-fitted to make secure connections with other devices. 2-1 heat-shrink ratio. 2012 Zeus Industrial Products. All rights reserved. **Sold for research purposes only and not for human use**