Learn More
Perkin Elmer US LLC PTFE 2 mL Weighing Cup for Standard or High Pressure Digestion Vessel, Qty. 16
Catalog No. 502111725
Encompass_Preferred
Weighing Cup for Standard 75 mL (40 Bar) Digestion Vessel or High Pressure 100 mL (100 Bar) Digestion Vessel for Titan MPS 8 and 16-Position Microwave Sample Preparation Systems.
Catalog No. 50-211-1725 Supplier Perkin Elmer US LLC Supplier No. N3134010
Weighing Cup for Standard 75 mL (40 Bar) Digestion Vessel or High Pressure 100 mL (100 Bar) Digestion Vessel for Titan MPS 8 and 16-Position Microwave Sample Preparation Systems.