Learn More
AVANTOR MASTERFLEX (VWR LLC) PTFE 2-WAY STOPCOCK, 1/8"" ORIF
Catalog No. NC2291323
Encompass
Masterflex PTFE 2-Way Stopcock, 1/8" Orifice, 1/4" OD Tube, 2" Length
This item is not returnable. View return policy
Catalog No. NC2291323 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9851572
Masterflex PTFE 2-Way Stopcock, 1/8" Orifice, 1/4" OD Tube, 2" Length