Learn More
Ace Glass, Inc. PTFE connecting adapter, 15mm Ace-Thred to 15-425 GPI vial, FETFE O-ring
SDP
Catalog No. 50983484
Encompass
PTFE connecting adapter, 15mm Ace-Thred to 15-425 GPI vial, FETFE O-ring
Catalog No. 50-983-484 Supplier Ace Glass, Inc. Supplier No. 1329015
PTFE connecting adapter, 15mm Ace-Thred to 15-425 GPI vial, FETFE O-ring