Learn More
Ace Glass, Inc. PTFE connecting adapter, 15mm Ace-Thred to 18-425 GPI vial, FETFE O-ring
SDP
Catalog No. 50977937
Encompass
PTFE connecting adapter, 15mm Ace-Thred to 18-425 GPI vial, FETFE O-ring
Catalog No. 50-977-937 Supplier Ace Glass, Inc. Supplier No. 1329018
PTFE connecting adapter, 15mm Ace-Thred to 18-425 GPI vial, FETFE O-ring