Learn More
Ace Glass, Inc. Adapter, #50 Ace-Thred™; to 3/4in FNPT, 59/64indrill-thru, PTFE
SDP
Catalog No. 50978512
Encompass_Preferred
Adapter, #50 Ace-Thred™; to 3/4in FNPT, 59/64indrill-thru, PTFE
Catalog No. 50-978-512 Supplier Ace Glass, Inc. Supplier No. 584497
Adapter, #50 Ace-Thred™; to 3/4in FNPT, 59/64indrill-thru, PTFE