Learn More
Analytical Sales & Services PTFE Adapter for Gray PEEK Ports on JustRite Waste Cans, for 3/16” Tubing
Catalog No. 5000805200
Encompass
PTFE Adapter for Gray PEEK Ports on JustRite Waste Cans, 3 Ports for 3/16” Tubing, 1 Port for Barbed Connector
Catalog No. 50-008-05200 Supplier Analytical Sales & Services Supplier No. CJR03
PTFE Adapter for Gray PEEK Ports on JustRite Waste Cans, 3 Ports for 3/16” Tubing, 1 Port for Barbed Connector