Learn More
Heidolph™ PTFE Adapter for Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900962 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-962 Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-962 Supplier Heidolph™ Supplier No. 036290034
Add to Cart
Add to Cart
PTFE adapter connects the PTFE seal and evaporating flask.
Included with Hei-VAP Industrial Rotary Evaporator
Specifications
Type Seal
For Use With (Equipment) Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov