Learn More
Ace Glass, Inc. PTFE baffle, 25.4mm diax250mm length shaft. Fits #25Ace-Thred adapter. Baffle dimensions: 32mmx360mmx14mm, 610mm overall length.
SDP
Catalog No. 50977544
Encompass
32mm X 360mm X 14mm PTFE Baffle. Overall length: 610mm. Use with #25 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.
Catalog No. 50-977-544 Supplier Ace Glass, Inc. Supplier No. 12177157
32mm X 360mm X 14mm PTFE Baffle. Overall length: 610mm. Use with #25 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.