Learn More
Ace Glass, Inc. PTFE baffle, 6.4mm diax175mm length shaft. Fits #7Ace-Thred adapter. Baffle dimensions-18mmx152mmx10mm, 327mm overall length.
SDP
Catalog No. 50977542
Encompass
18mm X 152mm X 10mm PTFE Baffle. Overall length: 327mm. Use with #7 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.
Catalog No. 50-977-542 Supplier Ace Glass, Inc. Supplier No. 12177145
18mm X 152mm X 10mm PTFE Baffle. Overall length: 327mm. Use with #7 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.