Learn More
Chemglass Life Sciences PTFE Baffle Cage, 4-Blade, 150mm ID Flange, for 2, 000mL Vessel
SDP
Catalog No. 501979406
Encompass_Preferred
PTFE insertable 4-blade baffle cage system for CG-1929-X series 100 and 150mm reactor systems. Baffle assembly has 4ea PTFE rectangular baffles attached 90 ° apart to a 100 or 150mm flange ring. Flange ring is inserted between the reaction vessel lid and reaction vessel. PTFE flange ring has an o-ring groove to accept a CG-147 O-Ring. The two and five liter sizes include a lower ring for stability. Please Note: CG-147 Perfluoro O-Rings are recommended for use with the baffle assembly.
Catalog No. 50-197-9406 Supplier Chemglass Life Sciences Supplier No. CG1971X2000
PTFE insertable 4-blade baffle cage system for CG-1929-X series 100 and 150mm reactor systems. Baffle assembly has 4ea PTFE rectangular baffles attached 90 ° apart to a 100 or 150mm flange ring. Flange ring is inserted between the reaction vessel lid and reaction vessel. PTFE flange ring has an o-ring groove to accept a CG-147 O-Ring. The two and five liter sizes include a lower ring for stability. Please Note: CG-147 Perfluoro O-Rings are recommended for use with the baffle assembly.