Learn More
Electron Microscopy Sciences PTFE Beaker, 2000 ml, 200 mm height, 114 mm diameter
SDP
Catalog No. 50996314
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 2000 ml
Height: 200 mm
Diameter: 114 mm
Catalog No. 50-996-314 Supplier Electron Microscopy Sciences Supplier No. 6094920
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 2000 ml
Height: 200 mm
Diameter: 114 mm