Learn More
Electron Microscopy Sciences PTFE Beaker, 600 ml, 127 mm height, 80 mm diameter
SDP
Catalog No. 501898981
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 600 ml
Height: 127 mm
Diameter: 80 mm
Catalog No. 50-189-8981 Supplier Electron Microscopy Sciences Supplier No. 6094906
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 600 ml
Height: 127 mm
Diameter: 80 mm