Learn More
Electron Microscopy Sciences PTFE Beaker Cap, 100 ml
SDP
Catalog No. 50996303
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 100 ml
Catalog No. 50-996-303 Supplier Electron Microscopy Sciences Supplier No. 60944C
You must be signed in and have a business account to purchase online.
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 100 ml