Learn More
Electron Microscopy Sciences PTFE Beaker Cap, 250 ml
SDP
Catalog No. 50996305
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 250 ml
Catalog No. 50-996-305 Supplier Electron Microscopy Sciences Supplier No. 60946C
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 250 ml