Learn More
Electron Microscopy Sciences PTFE Beakers, 1 ml, 18 mm height, 10 mm diameter
SDP
Catalog No. 50996292
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 1 ml
Height: 18 mm
Diameter: 10 mm
Catalog No. 50-996-292 Supplier Electron Microscopy Sciences Supplier No. 60936
Add to Cart
Add to Cart
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 1 ml
Height: 18 mm
Diameter: 10 mm