Learn More
Electron Microscopy Sciences PTFE Beakers, 400 ml, 106 mm height, 73 mm diameter
SDP
Catalog No. 501898979
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 400 ml
Height: 106 mm
Diameter: 73 mm
Catalog No. 50-189-8979 Supplier Electron Microscopy Sciences Supplier No. 60947
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 400 ml
Height: 106 mm
Diameter: 73 mm