Learn More
Electron Microscopy Sciences PTFE Beakers, 50 ml, 58 mm height, 38 mm diameter
SDP
Catalog No. 50996300
Encompass_Preferred
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 50 ml
Height: 58 mm
Diameter: 38 mm
Catalog No. 50-996-300 Supplier Electron Microscopy Sciences Supplier No. 60942
Is statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination. Supplied with a pouring spout and they are virtually immune to chemical attack. Working temperature range is from cryogenic to 260 degree C (500 degree F). Ideal for use in wafer etching.
Capacity: 50 ml
Height: 58 mm
Diameter: 38 mm