Learn More
Ace Glass, Inc. PTFE bearing, 24/40 inner joint, 10mm shaft
SDP
Catalog No. NC9355826
Encompass
Debris-free PTFE stirrer bearing with 24/40 inner joint, for use with 10mm shaft, totally eliminates flaking into the reaction. Vacuum tight and chemically resistant. 500rpm maximum with polished glass or stainless steel stir shafts.
Catalog No. NC9355826 Supplier Ace Glass, Inc. Supplier No. 1344530
Debris-free PTFE stirrer bearing with 24/40 inner joint, for use with 10mm shaft, totally eliminates flaking into the reaction. Vacuum tight and chemically resistant. 500rpm maximum with polished glass or stainless steel stir shafts.