Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PTFE Bottles

Catalog No. AA38111KT
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1 L
2000 mL
Height (Metric):
61 mm
76 mm
88 mm
115 mm
150 mm
185 mm
240 mm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Height (Metric)
AA38111KT 50 mL 76 mm
AA38112KT 100 mL 88 mm
AA38113KT 250 mL 115 mm
AA38114KT 500 mL 150 mm
AA38115KT 1 L 185 mm
AA38110KT 25 mL 61 mm
AA38116KT 2000 mL 240 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. AA38111KT Supplier Thermo Scientific™ Supplier No. 38111KT
Add to Cart

For use with aggressive or valuable reagents

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Inert and leak-free
  • Flexible and with PTFE cap
TRUSTED_SUSTAINABILITY

Specifications

Capacity 50 mL
Capacity (Metric) 50 mL
Material PTFE
Content And Storage Ambient temperatures
Height (Metric) 76 mm
Product Type Reagent Bottle
Diameter (Metric) 43 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.