Learn More
Perkin Elmer US LLC Un-Assembled Aluminum Crimp Cap with PTFE/Silicone (White) Septa and Star Spring, 100/pk
Catalog No. 509051862
Encompass_Preferred
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.
Catalog No. 50-905-1862 Supplier Perkin Elmer US LLC Supplier No. B0104241
For sealing sample vials for temperatures up to 210°C. Recommend for storage and for multiple injection use.