Learn More
Electron Microscopy Sciences PTFE Coated Tip Forceps
SDP
Catalog No. 5027484
Encompass_Preferred
Nickel-plated forceps with "PTFE" coated tips. Offers a soft and good grip to handle delicate articles. Straight, "PTFE" coated tips, corrosion-free. Offers a soft and good grip to handle delicate articles. Overall length 37/8"(98mm), medium points.
Catalog No. 50-274-84 Supplier Electron Microscopy Sciences Supplier No. 72973
Nickel-plated forceps with "PTFE" coated tips. Offers a soft and good grip to handle delicate articles. Straight, "PTFE" coated tips, corrosion-free. Offers a soft and good grip to handle delicate articles. Overall length 37/8"(98mm), medium points.