Learn More
Glass Expansion PTFE Coated Viton O-rings (PKT 10)
SDP
Catalog No. 502702921
Encompass_Preferred
PTFE Coated Viton O-Rings (PKT 10). The O-Ring removal tool (50-100-1636) facilitates the removal and installation of O-rings used with Glass Expansion D-Torches. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2921 Supplier Glass Expansion Supplier No. VT007
PTFE Coated Viton O-Rings (PKT 10). The O-Ring removal tool (50-100-1636) facilitates the removal and installation of O-rings used with Glass Expansion D-Torches. This tool is especially handy while wearing PPE that can limit dexterity.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001