Learn More
Cole-Parmer Diba Omnifit PTFE Cone (Ferrule) for Connector and Valve Caps, 1/16" (1.6 mm); 4/pk
Catalog No. NC1783968 Shop All Cole-Parmer Products
Encompass
Select PTFE cones when chemical resistance is important. The OD of the tubing should be matched to the inside diameter of the cone.
Catalog No. NC1783968 Supplier Cole-Parmer Supplier No. 2193947
Select PTFE cones when chemical resistance is important. The OD of the tubing should be matched to the inside diameter of the cone.