Learn More
Ace Glass, Inc. Instatherm; oil bath, 2600mL, 182mm ID, 190mm OD, 100mm height, 120v/10amp max, 12ft PTFE-clad, 2-prong, permanently attached controller cord, thermocouple/thermometer holding clip
SDP
Catalog No. 5097775
Encompass
Instatherm; oil bath, 2600mL, 182mm ID, 190mm OD, 100mm height, 120v/10amp max, 12ft PTFE-clad, 2-prong, permanently attached controller cord, thermocouple/thermometer holding clip
Catalog No. 50-977-75 Supplier Ace Glass, Inc. Supplier No. 960247
Instatherm; oil bath, 2600mL, 182mm ID, 190mm OD, 100mm height, 120v/10amp max, 12ft PTFE-clad, 2-prong, permanently attached controller cord, thermocouple/thermometer holding clip