Learn More
Ace Glass, Inc. Instatherm; oil bath, 700mL, 119mm ID, 125mm OD, 65mm height, 40v/8amp max, 12ft PTFE-clad, 2-prong, permanently attached controller cord, thermocouple/thermometer holding clip
SDP
Catalog No. 5097773
Encompass
Instatherm; oil bath, 700mL, 119mm ID, 125mm OD, 65mm height, 40v/8amp max, 12ft PTFE-clad, 2-prong, permanently attached controller cord, thermocouple/thermometer holding clip
Catalog No. 50-977-73 Supplier Ace Glass, Inc. Supplier No. 960243
Instatherm; oil bath, 700mL, 119mm ID, 125mm OD, 65mm height, 40v/8amp max, 12ft PTFE-clad, 2-prong, permanently attached controller cord, thermocouple/thermometer holding clip