Learn More
Electron Microscopy Sciences PTFE Crucibles, 25 ml
SDP
Catalog No. 5032934
Encompass_Preferred
Manufactured from pure "PTFE", chemically inert, and usable to 280 degree C.
Volume: 25 ml
Catalog No. 50-329-34 Supplier Electron Microscopy Sciences Supplier No. 6650925
Manufactured from pure "PTFE", chemically inert, and usable to 280 degree C.
Volume: 25 ml