Learn More
Electron Microscopy Sciences PTFE Crucibles, 5 ml
SDP
Catalog No. 5032936
Encompass_Preferred
Manufactured from pure "PTFE", chemically inert, and usable to 280 degree C.
Volume: 5 ml
Catalog No. 50-329-36 Supplier Electron Microscopy Sciences Supplier No. 665095
Manufactured from pure "PTFE", chemically inert, and usable to 280 degree C.
Volume: 5 ml