Learn More
Electron Microscopy Sciences PTFE Crucibles, 75 ml
SDP
Supplier:  Electron Microscopy Sciences 6650975
Encompass_Preferred
Manufactured from pure "PTFE", chemically inert, and usable to 280 degree C.
Volume: 75 ml
Catalog No. 50-329-37