Learn More
Bellco Glass, Inc. Teflon Cushion 38 mm
SDP
Catalog No. 501944655
Encompass_Preferred
Stabilizes pipette canister during use. PIPETE CANISTER CUSHION, PTFE. CAN BE REPEATEDLY STERILIZED USING DRY OR STEAM HEAT COMPLETELY NON-TOXIC, LINT FREE, PERMANENTLY RESILIENT 38MM CUSHIONS FIT 1287 SERIES ROUND BOTTOM, GLASS PIPETTE CANISTERS 63MM ROUND CUSHIONS FIT OTHER MANUFACTURERS' ROUND, STEEL CANISTERS. Qty: 1 ea.
Catalog No. 50-194-4655 Supplier Bellco Glass, Inc. Supplier No. 128688815
Stabilizes pipette canister during use. PIPETE CANISTER CUSHION, PTFE. CAN BE REPEATEDLY STERILIZED USING DRY OR STEAM HEAT COMPLETELY NON-TOXIC, LINT FREE, PERMANENTLY RESILIENT 38MM CUSHIONS FIT 1287 SERIES ROUND BOTTOM, GLASS PIPETTE CANISTERS 63MM ROUND CUSHIONS FIT OTHER MANUFACTURERS' ROUND, STEEL CANISTERS. Qty: 1 ea.