Learn More
Chemglass Life Sciences Diaphragm Liquid Metering Pump, 6L/Min
SDP
Catalog No. 501531032
Encompass_Preferred
Diaphragm Liquid Metering Pump, 6L/Min. Specifications: Dimensions H x W x D: 10 x 6.5 x 4.5. Pump Head: PVDF. Diaphragm: PTFE Coated. Valves: FFPM. Maximum Head Pressure: 14.5 psi. Power Requirements: 115V, 60Hz. Head Connections: 1/2 Compression Fittings. PLEASE NOTE: DESIGNED FOR SCRUBBER STAND MOUNTING, NOT A BENCHTOP MODEL.
Catalog No. 50-153-1032 Supplier Chemglass Life Sciences Supplier No. CG117150
Diaphragm Liquid Metering Pump, 6L/Min. Specifications: Dimensions H x W x D: 10 x 6.5 x 4.5. Pump Head: PVDF. Diaphragm: PTFE Coated. Valves: FFPM. Maximum Head Pressure: 14.5 psi. Power Requirements: 115V, 60Hz. Head Connections: 1/2 Compression Fittings. PLEASE NOTE: DESIGNED FOR SCRUBBER STAND MOUNTING, NOT A BENCHTOP MODEL.