Learn More
Perkin Elmer US LLC PTFE Digestion Tube 50 mL, Includes Blue Caps
Catalog No. 502103727
Encompass_Preferred
All-in-one tubes eliminate the need for five separate vessels: Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.
Catalog No. 50-210-3727 Supplier Perkin Elmer US LLC Supplier No. N9308024
All-in-one tubes eliminate the need for five separate vessels: Digestion beakers, volumetric flasks, graduated cylinders, autosampler tubes and storage containers.