Learn More
Description
- Ideal for single use applications mixing radioactive reagents or when cross contamination might be a concern.
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert.
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Specifications
Specifications
| Quantity | 100 |
| Shape | Cylinder |
| Certifications/Compliance | ISO 9001:2008 |
| Autoclavable | Autoclavable |
| Diameter (English) | 0.31 in. |
| Length (English) | 1.61 in. |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 41.3 mm |
| Coating | PTFE |
| Color | White |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.