Learn More
Chemglass Life Sciences Porous PTFE Filter Disc, 19mm & 25.4mm
SDP
Catalog No. CG209331
Encompass_Preferred
Porous PTFE Filter Disc, 19mm & 25.4mm. 50 micron porosity PTFE filter disc. Replacement part for CG2093 series of PTFE Flake Retaining Cup.
Catalog No. CG209331 Supplier Chemglass Life Sciences Supplier No. CG209331
Porous PTFE Filter Disc, 19mm & 25.4mm. 50 micron porosity PTFE filter disc. Replacement part for CG2093 series of PTFE Flake Retaining Cup.