Learn More
Chemglass Life Sciences 100mm Envelope Style Gasket
SDP
Catalog No. 50872160
Encompass_Preferred
100mm Envelope Style Gasket. These envelope style gaskets are machined using chemically resistant virgin PTFE and have a silicone core. The larger sealing surface results in improved sealing under full vacuum, as opposed to other o-ring materials. The machined in o-ring aids in sealing and centering of gasket on the Duran o-ring flange.
Catalog No. 50-872-160 Supplier Chemglass Life Sciences Supplier No. CG147G03
100mm Envelope Style Gasket. These envelope style gaskets are machined using chemically resistant virgin PTFE and have a silicone core. The larger sealing surface results in improved sealing under full vacuum, as opposed to other o-ring materials. The machined in o-ring aids in sealing and centering of gasket on the Duran o-ring flange.