Learn More
Cole-Parmer Cole-Parmer 21-3S Fiberglass Tape with PTFE Coating, 1" W, 5 mil thick; 18 yd/Roll
Catalog No. NC9080907 Shop All Cole-Parmer Products
Encompass_Preferred
Tapes are made from woven fiberglass fabric and have silicone adhesive on one side. Tapes have excellent chemical, heat, and moisture resistance. Use for packaging, heat sealing, or shrink wrapping. Approved by USDA and is FDA compliant for food contact. Withstand temperatures from –100 to 500°F (–73 to 260 C).
Catalog No. NC9080907 Supplier Cole-Parmer Supplier No. 0653204
Tapes are made from woven fiberglass fabric and have silicone adhesive on one side. Tapes have excellent chemical, heat, and moisture resistance. Use for packaging, heat sealing, or shrink wrapping. Approved by USDA and is FDA compliant for food contact. Withstand temperatures from –100 to 500°F (–73 to 260 C).