Learn More
IW Tremont Co Inc LabExact PTFE inline membrane filter, 50mm dia, porosity 0.22um, STERILE filter with 1/4"" to 3/8 barb ports, 10 pack
SDP
Supplier:  IW Tremont Co Inc LEIWTES10500
Encompass_Preferred
LabExact PTFE inline membrane filter, 50mm dia, porosity 0.22um, STERILE filter with 1/4" to 3/8¿ barb ports, 10 pack
Catalog No. 50-223-7851