Learn More
Ace Glass, Inc. PTFE filter reactor, 250mL, complete, features (4)24/40 head and #50 valved bottom filter assembly
SDP
Supplier:  Ace Glass, Inc. 639002
Encompass
250mL PTFE filter reactor with polypropylene filter support. Reactor head features four 24/40 joints. #50 valved bottom filter assembly facilitates easy filtrate recovery and cleaning. Reactor is suited for fluorine chemistry work as well as any work using strong alkalis where glass cannot be used.
Catalog No. 50-972-824