Learn More
Chemglass Life Sciences 19mm PTFE Flake Retaining Cup
SDP
Catalog No. CG209320
Encompass_Preferred
19mm PTFE Flake Retaining Cup. Cup is designed to catch all flaking that occurs with PTFE stirrer bearings. Made entirely of PTFE, the complete item consists of a compression fitting, deep perforated PTFE cup and a 50 micron porosity PTFE filter disc.
Catalog No. CG209320 Supplier Chemglass Life Sciences Supplier No. CG209320
19mm PTFE Flake Retaining Cup. Cup is designed to catch all flaking that occurs with PTFE stirrer bearings. Made entirely of PTFE, the complete item consists of a compression fitting, deep perforated PTFE cup and a 50 micron porosity PTFE filter disc.