Learn More
Perkin Elmer US LLC PTFE Flame Sampling Probe Assembly with Carbon Fiber Support and Male Screw Fitting, 0.55 mm ID for AA/ICP/ICPMS
Catalog No. 502112252
Encompass_Preferred
For ICP/ICPMS requires but does not include N8145018*. For AA with a stainless steel nebulizer, the male connect should be removed so that tubing can be directly connected to the nebulizer, no additional tubing is required.
Catalog No. 50-211-2252 Supplier Perkin Elmer US LLC Supplier No. N2024132
Add to Cart
Add to Cart
For ICP/ICPMS requires but does not include N8145018*. For AA with a stainless steel nebulizer, the male connect should be removed so that tubing can be directly connected to the nebulizer, no additional tubing is required.