Learn More
Neta Scientific Agilent PTFE Frits 5PK
SDP
Catalog No. 50925595
Encompass_Preferred
This item is manufactured by Agilent
Agilent PTFE frits, 5/pk. For 1290 Infinity Binary Pumps (G4220A/B)and all 1260 Infinity/1200 Series/1100 Series analytical pumps) - Purge Valves
UNSPSC Code  41105106
- AA
Catalog No. 50-925595 Supplier Neta Scientific Supplier No. 0101822707
This item is manufactured by Agilent
Agilent PTFE frits, 5/pk. For 1290 Infinity Binary Pumps (G4220A/B)and all 1260 Infinity/1200 Series/1100 Series analytical pumps) - Purge Valves
UNSPSC Code  41105106
- AA