Learn More
Chemglass Life Sciences 10mm PTFE Gasket
SDP
Catalog No. CG204901
Encompass_Preferred
10mm PTFE Gasket. PTFE gasket is 0.015" thick with an opening to fit Chemglass stirrer shafts. Gasket is placed below the CG-2048 Safety collar to prevent wear as a result of the collar rotating against the stirrer bearing. Also useful as a dust cover. SOLD 12/PKG.
Catalog No. CG204901 Supplier Chemglass Life Sciences Supplier No. CG204901
10mm PTFE Gasket. PTFE gasket is 0.015" thick with an opening to fit Chemglass stirrer shafts. Gasket is placed below the CG-2048 Safety collar to prevent wear as a result of the collar rotating against the stirrer bearing. Also useful as a dust cover. SOLD 12/PKG.