Learn More
Curiox Biosystems Inc PTFE GRID FOR DIRECT READING O
SDP
Catalog No. NC2459468
Encompass
  • Volume adaptor designed for enhanced sample preparation workflows.
  • Constructed from PTFE material.
  • Compatible with Curiox Laminar Wash 96-well plates.
  • Increases well capacity up to 300 µL.
  • Facilitates direct acquisition of samples for flow cytometry.
  • Enables automation, improving workflow efficiency.
  • Optimized for consistent and reproducible data.
  • Available in manual and robotic gripper formats.
  • Validated for use with multiple flow cytometers.
  • Disposable for single-use only.
  • Not intended for reagent incubation.
Catalog No. NC2459468 Supplier Curiox Biosystems Inc Supplier No. GRPF9601202308220922
  • Volume adaptor designed for enhanced sample preparation workflows.
  • Constructed from PTFE material.
  • Compatible with Curiox Laminar Wash 96-well plates.
  • Increases well capacity up to 300 µL.
  • Facilitates direct acquisition of samples for flow cytometry.
  • Enables automation, improving workflow efficiency.
  • Optimized for consistent and reproducible data.
  • Available in manual and robotic gripper formats.
  • Validated for use with multiple flow cytometers.
  • Disposable for single-use only.
  • Not intended for reagent incubation.