Learn More
BURKLE INC PTFE hose, Ø 0.24x0.31 in. (6x8 mm), pressure max. 580.15 psi. (40 bar), 16.40 ft. (5 m)
Catalog No. 5000026083
Encompass_Preferred
PTFE hoses have outstanding properties, they are extremely resistant to almost all chemicals, have the lowest coefficient of friction of all polymers and excellent non-stick/residual emptying properties. PTFE hoses do not smell, burn or smoke and are resistant to ageing and UV radiation. However, they are not resistant to fluorine compounds and strong reducing agents.
PTFE tubing has good transparency for excellent visual control and excellent abrasion and aging resistance, which ensures long-term reliability.
Catalog No. 50-000-26083 Supplier BURKLE INC Supplier No. 88040608
PTFE hoses have outstanding properties, they are extremely resistant to almost all chemicals, have the lowest coefficient of friction of all polymers and excellent non-stick/residual emptying properties. PTFE hoses do not smell, burn or smoke and are resistant to ageing and UV radiation. However, they are not resistant to fluorine compounds and strong reducing agents.
PTFE tubing has good transparency for excellent visual control and excellent abrasion and aging resistance, which ensures long-term reliability.