Learn More
Chemglass Life Sciences PTFE Inner Bearing with O-Ring, 25.4mm
SDP
Catalog No. CG196876
Encompass_Preferred
PTFE Inner Bearing with O-Ring, 25.4mm. Grease-Free PTFE stirrer bearing designed for use in moderate vacuum applications. Bearing consists of a threaded glass adapter having a 45/50 inner joint, a saddle o-ring, a bushing with compression and locking nut, and a precision PTFE inner bearing. The saddle o-ring provides only glass and PTFE contact with the reaction, while allowing for the vacuum tight seal. Design of the bearing permits replacement of the saddle o-ring at a fraction of the cost of the entire bearing.
Catalog No. CG196876 Supplier Chemglass Life Sciences Supplier No. CG196876
PTFE Inner Bearing with O-Ring, 25.4mm. Grease-Free PTFE stirrer bearing designed for use in moderate vacuum applications. Bearing consists of a threaded glass adapter having a 45/50 inner joint, a saddle o-ring, a bushing with compression and locking nut, and a precision PTFE inner bearing. The saddle o-ring provides only glass and PTFE contact with the reaction, while allowing for the vacuum tight seal. Design of the bearing permits replacement of the saddle o-ring at a fraction of the cost of the entire bearing.